ENAH38A2D8B6 Enamine

methyl (2R)-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoate hydrochloride

  • MDL number MFCD31915846
  • Cas Number : 2309431-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764296
Availability details for
Quantity
  • Availability
  • Add to Cart