ENAH38A2D8BC Enamine

1-(2-{[(tert-butoxy)carbonyl]amino}ethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD31915558
  • Cas Number : 2309457-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764324
Availability details for
Quantity
  • Availability
  • Add to Cart