ENAH38A2D8BD Enamine

6-(4-fluorophenyl)-4H,5H-[1,2,3]triazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD31915847
  • Cas Number : 2309449-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764331
Availability details for
Quantity
  • Availability
  • Add to Cart