ENAH38A2D8C3 Enamine

sodium dimethyl sulfanidylphosphonate

  • MDL number MFCD28404549
  • Cas Number : 23754-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764351
Availability details for
Quantity
  • Availability
  • Add to Cart