ENAH38A2D8C6 Enamine

1-[cyclopropyl(2-hydroxypropyl)amino]propan-2-ol

  • MDL number MFCD19313580
  • Cas Number : 1267109-37-4
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764378
Availability details for
Quantity
  • Availability
  • Add to Cart