ENAH38A2D8CA Enamine

6-amino-3-methyl-3,4-dihydro-2H-1,3-benzoxazin-2-one hydrochloride

  • MDL number MFCD31915864
  • Cas Number : 2309474-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764409
Availability details for
Quantity
  • Availability
  • Add to Cart