ENAH38A2D8D2 Enamine

5-oxaspiro[3.4]octan-6-one

  • MDL number MFCD31915694
  • Cas Number : 211311-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764452
Availability details for
Quantity
  • Availability
  • Add to Cart