ENAH38A2D8DA Enamine

1-amino-3-tert-butylcyclobutane-1-carboxylic acid hydrochloride

  • MDL number MFCD31915871
  • Cas Number : 2309473-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764494
Availability details for
Quantity
  • Availability
  • Add to Cart