ENAH38A2D8DC Enamine

{1,4-dioxaspiro[4.6]undecan-7-yl}methanamine

  • MDL number MFCD20707733
  • Cas Number : 38745-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764499
Availability details for
Quantity
  • Availability
  • Add to Cart