ENAH38A2D8DD Enamine

(3,3-difluorocycloheptyl)methanol

  • MDL number MFCD31915872
  • Cas Number : 2309465-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764500
Availability details for
Quantity
  • Availability
  • Add to Cart