ENAH38A2D8E4 Enamine

(1s,3s)-3-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}-3-hydroxycyclobutane-1-carboxylic acid

  • MDL number MFCD31915747
  • Cas Number : 2309468-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764524
Availability details for
Quantity
  • Availability
  • Add to Cart