ENAH38A2D8FC Enamine

4-bromo-3,3-dimethylbut-1-yne

  • MDL number MFCD31915759
  • Cas Number : 2309463-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764776
Availability details for
Quantity
  • Availability
  • Add to Cart