ENAH38A2D902 Enamine

methyl 3-(2-aminopropan-2-yl)-1H-pyrazole-5-carboxylate hydrochloride

  • MDL number MFCD31915762

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764793
Availability details for
Quantity
  • Availability
  • Add to Cart