ENAH38A2D91A Enamine

(1r,4r)-4-(1H-1,2,4-triazol-3-yl)cyclohexan-1-amine hydrochloride

  • MDL number MFCD31915889
  • Cas Number : 2309458-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764920
Availability details for
Quantity
  • Availability
  • Add to Cart