ENAH38A2D91B Enamine

ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazolo[4,3-b]pyridine-6-carboxylate

  • MDL number MFCD31915818
  • Cas Number : 2309463-07-6
  • Molecular Weight : 273.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764939
Availability details for
Quantity
  • Availability
  • Add to Cart