ENAH38A2D928 Enamine

[(1R,2R,4S,5S)-4-aminobicyclo[3.1.0]hexan-2-yl]methanol hydrochloride

  • MDL number MFCD31915121
  • Cas Number : 2309431-91-0
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766461
Availability details for
Quantity
  • Availability
  • Add to Cart