ENAH38A2D92A Enamine

5-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}azetidin-3-yl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD31915123
  • Cas Number : 2309457-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766474
Availability details for
Quantity
  • Availability
  • Add to Cart