ENAH38A2D92B Enamine

3-(ethylamino)propanamide hydrochloride

  • MDL number MFCD29924219
  • Cas Number : 2044705-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766477
Availability details for
Quantity
  • Availability
  • Add to Cart