ENAH38A2D934 Enamine

3-[(piperidin-3-yl)methyl]phenol hydrobromide

  • MDL number MFCD31915125
  • Cas Number : 2309445-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766556
Availability details for
Quantity
  • Availability
  • Add to Cart