ENAH38A2D938 Enamine

2-(4-methyl-3-nitrophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD31915127
  • Cas Number : 2309459-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766592
Availability details for
Quantity
  • Availability
  • Add to Cart