ENAH38A2D939 Enamine

methyl 3-amino-2-[(4-chlorophenyl)methyl]propanoate hydrochloride

  • MDL number MFCD31915581
  • Cas Number : 2309454-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766596
Availability details for
Quantity
  • Availability
  • Add to Cart