ENAH38A2D93E Enamine

1-hydroxy-4-phenylcyclohexane-1-carboxylic acid

  • MDL number MFCD04971281
  • Cas Number : 1248776-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766642
Availability details for
Quantity
  • Availability
  • Add to Cart