ENAH38A2D94A Enamine

ethyl 2-(aminomethyl)-4-(difluoromethyl)pyrimidine-5-carboxylate hydrochloride

  • MDL number MFCD31915588
  • Cas Number : 2309454-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766748
Availability details for
Quantity
  • Availability
  • Add to Cart