ENAH38A2D952 Enamine

sodium 4-amino-1-methyl-1H-imidazole-5-carboxylate

  • MDL number MFCD31915650
  • Cas Number : 2309461-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766798
Availability details for
Quantity
  • Availability
  • Add to Cart