ENAH38A2D954 Enamine

6-bromopyridine-2-carboximidamide hydrochloride

  • MDL number MFCD12755610
  • Cas Number : 1245806-87-4
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766802
Availability details for
Quantity
  • Availability
  • Add to Cart