ENAH38A2D955 Enamine

5-(piperidin-4-yl)-1H-pyrazole-3-carboxylic acid hydrochloride

  • MDL number MFCD31915651

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766803
Availability details for
Quantity
  • Availability
  • Add to Cart