ENAH38A2D957 Enamine

[(4-chloro-3-methylphenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD31915652
  • Cas Number : 2309468-72-0
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766810
Availability details for
Quantity
  • Availability
  • Add to Cart