ENAH38A2D95A Enamine

4-[(4-methylphenyl)amino]pyrimidine-5-carboxylic acid

  • MDL number MFCD11573883
  • Cas Number : 16100-48-4
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766822
Availability details for
Quantity
  • Availability
  • Add to Cart