ENAH38A2D95F Enamine

3-tert-butyl-4-methylbenzoic acid

  • MDL number MFCD28356927
  • Cas Number : 1369784-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766911
Availability details for
Quantity
  • Availability
  • Add to Cart