ENAH38A2D962 Enamine

4-[(3-bromophenyl)amino]-2-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD26661485
  • Cas Number : 1542002-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766915
Availability details for
Quantity
  • Availability
  • Add to Cart