ENAH38A2D963 Enamine

4-[(3-fluorophenyl)amino]-2-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD26655853
  • Cas Number : 1529588-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766916
Availability details for
Quantity
  • Availability
  • Add to Cart