ENAH38A2D964 Enamine

2-methoxy-3,3-dimethylcyclobutan-1-amine

  • MDL number MFCD31915136
  • Cas Number : 2309456-64-0
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766919
Availability details for
Quantity
  • Availability
  • Add to Cart