ENAH38A2D966 Enamine

methyl (5r,8r)-1-azaspiro[4.5]decane-8-carboxylate hydrochloride

  • MDL number MFCD31915137
  • Cas Number : 2309462-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6766924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6766924
Availability details for
Quantity
  • Availability
  • Add to Cart