ENAH38A2D974 Enamine

lithium(1+) ion 4-(1H-1,2,4-triazol-3-yl)butanoate

  • MDL number MFCD31915665
  • Cas Number : 2309446-45-3
  • Molecular Weight : 161.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6767013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6767013
Availability details for
Quantity
  • Availability
  • Add to Cart