ENAH38A2D987 Enamine

methyl (2R)-2-(2-aminoacetamido)-3-methylbutanoate hydrochloride

  • MDL number MFCD31915185
  • Cas Number : 857506-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6767159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6767159
Availability details for
Quantity
  • Availability
  • Add to Cart