ENAH38A2D98B Enamine

azecan-2-one

  • MDL number MFCD00154291
  • Cas Number : 6142-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6770961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6770961
Availability details for
Quantity
  • Availability
  • Add to Cart