ENAH38A2D98E Enamine

5-amino-3-(propan-2-yl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD20621004
  • Cas Number : 1366472-78-7
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6773892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6773892
Availability details for
Quantity
  • Availability
  • Add to Cart