ENAH38A2D99D Enamine

5-oxaspiro[3.4]octane-1-carboxylic acid

  • MDL number MFCD31915857
  • Cas Number : 2309467-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6998911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6998911
Availability details for
Quantity
  • Availability
  • Add to Cart