ENAH38A2D99F Enamine

(3R)-3-amino-1-methylazepan-2-one

  • MDL number MFCD19689124
  • Cas Number : 1375245-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7045114

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
hcodes Xi,C
MSDS https://www.enaminestore.com/catalog/EN300-7045114
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart