ENAH38A2D9A2 Enamine

(5-methyl-4,5-dihydro-1,2-oxazol-3-yl)methanol

  • MDL number MFCD31915858
  • Cas Number : 1823333-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7063735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7063735
Availability details for
Quantity
  • Availability
  • Add to Cart