ENAH38A2D9A4 Enamine

(2E)-3-(cyclopent-1-en-1-yl)prop-2-enoic acid

  • MDL number MFCD03093333
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7096773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7096773
Availability details for
Quantity
  • Availability
  • Add to Cart