ENAH38A2D9AC Enamine

(2,2,2-trimethoxyethyl)cyclopropane

  • MDL number MFCD22681606
  • Cas Number : 1391407-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7147065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7147065
Availability details for
Quantity
  • Availability
  • Add to Cart