ENAH38A2D9AF Enamine

(oxiran-2-yl)methyl methanesulfonate

  • MDL number MFCD00128141
  • Cas Number : 6177-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7164827

Safety Information

signalword Danger
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
hcodes Carc Cat 3,Muta Cat 3,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-7164827
pictograms GHS05,GHS07,GHS08
hstatements H302,H312,H315,H318,H332,H335,H341,H351
Availability details for
Quantity
  • Availability
  • Add to Cart