ENAH38A2D9B0 Enamine

(2R)-2-cyclopentyl-2-hydroxyacetic acid

  • MDL number MFCD19231483
  • Cas Number : 174603-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7171097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7171097
Availability details for
Quantity
  • Availability
  • Add to Cart