ENAH38A2D9B9 Enamine

5-benzyl-5-azaspiro[3.5]nonan-8-ol

  • MDL number MFCD31915194
  • Cas Number : 2309465-95-8
  • Molecular Weight : 231.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7216656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7216656
Availability details for
Quantity
  • Availability
  • Add to Cart