ENAH38A2D9BA Enamine

N,N-dibenzylazetidin-3-amine

  • MDL number MFCD17385962
  • Cas Number : 931414-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7249647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7249647
Availability details for
Quantity
  • Availability
  • Add to Cart