ENAH38A2D9C2 Enamine

tert-butyl 3-(4-methoxy-3,4-dioxobutanoyl)azetidine-1-carboxylate

  • MDL number MFCD31915905
  • Cas Number : 2309463-98-5
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7351950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7351950
Availability details for
Quantity
  • Availability
  • Add to Cart