ENAH38A2D9C9 Enamine

4H,5H,6H-cyclopenta[c]thiophen-5-amine hydrochloride

  • MDL number MFCD31915908
  • Cas Number : 2309465-28-7
  • Molecular Weight : 175.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352030
Availability details for
Quantity
  • Availability
  • Add to Cart