ENAH38A2D9CA Enamine

methyl 2-({1-[(tert-butoxy)carbonyl]piperidin-4-yl}methoxy)pyridine-4-carboxylate

  • MDL number MFCD31915909
  • Cas Number : 2309456-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352033
Availability details for
Quantity
  • Availability
  • Add to Cart