ENAH38A2D9CC Enamine

[(4-amino-2,5-dimethoxyphenyl)sulfanyl]formonitrile

  • MDL number MFCD31915910
  • Cas Number : 2309465-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352041
Availability details for
Quantity
  • Availability
  • Add to Cart